Learn More
Cayman Chemical IsoImperatrIn 50mg
SDP
Catalog No. 502982471
Encompass_Preferred
A natural furanocoumarin that inhibits CYP1A activity, in particular blocking hepatic EROD activity; also induces the expression of GSTα by activating Nrf2
Catalog No. 50-298-2471 Supplier Cayman Chemical Supplier No. 1985150 MG
A natural furanocoumarin that inhibits CYP1A activity, in particular blocking hepatic EROD activity; also induces the expression of GSTα by activating Nrf2