Learn More
Cayman Chemical IsokotnIn B 2.5mg
SDP
Catalog No. 502933750
Encompass_Preferred
A fungal metabolite; dietary administration reduces C. hemipterus larvae feeding by 21% at 100 ppm
Catalog No. 50-293-3750 Supplier Cayman Chemical Supplier No. 250822.5 MG
A fungal metabolite; dietary administration reduces C. hemipterus larvae feeding by 21% at 100 ppm