Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mystaire™ Isola™ Edge Laboratory Fixtures
SDP

Catalog No. 15338933 Shop All Mystaire Products
Change view
Click to view available options
Product Type:
Air Fixture
Argon Gas Fixture
DI Water Fixture
Helium Gas Fixture
Natural Gas Fixture
Nitrogen Gas
Vacuum Fixture
Water Fixture
Description:
4 in.
45° Barb Connection
6 in.
Goose Neck Connection
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Description
15-338-933 DI Water Fixture Goose Neck Connection
15-338-927 Helium Gas Fixture 45° Barb Connection
15-338-928 Natural Gas Fixture 45° Barb Connection
15-338-929 Nitrogen Gas 45° Barb Connection
15-338-931 Water Fixture 4 in.
15-338-932 Water Fixture 6 in.
15-338-926 Water Fixture Goose Neck Connection
15-338-923 Air Fixture 45° Barb Connection
15-338-922 Argon Gas Fixture 45° Barb Connection
15-338-930 Vacuum Fixture 45° Barb Connection
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 15-338-933 Supplier Mystaire™ Supplier No. MYFXWTDI
Add to Cart
Add to Cart

The same conveniences found on traditional laboratory hoods now in a filtered workstation.

  • Wall mounted remote control
  • Main chamber connection

Specifications

For Use With (Equipment) Isola Edge Filtered Workstation
Description Goose Neck Connection
Product Type DI Water Fixture
Color Green
Configuration Options Wall Mounted
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.