Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isolariciresinol | 548-29-8 | 97.3% | C20H24O6 | 10MG
SDP

Catalog No. 5000259488
Encompass_Preferred

Isolariciresinol is a naturally occurring phenolic lignan used as a research-grade reference compound with reported anti-inflammatory activity. It is provided with documented purity and storage recommendations for use in biochemical and pharmacological studies.

  • High purity (≈97.3%).
  • Molecular formula C20H24O6.
  • Molecular weight 360.40 g/mol.
  • Available in small pack sizes, including 10 mg.
  • Storage: store at 4°C and protect from light; in solvent: -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-59488 Supplier Medchemexpress LLC Supplier No. HY1457910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isolariciresinol is a naturally occurring phenolic lignan used as a research-grade reference compound with reported anti-inflammatory activity. It is provided with documented purity and storage recommendations for use in biochemical and pharmacological studies.

  • High purity (≈97.3%).
  • Molecular formula C20H24O6.
  • Molecular weight 360.40 g/mol.
  • Available in small pack sizes, including 10 mg.
  • Storage: store at 4°C and protect from light; in solvent: -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.