Learn More
Cayman Chemical Isolariciresinol, Purity: ≥95%, CAS: 548-29-8, 5 mg
SDP
Catalog No. 502911977
Encompass_Preferred
A lignan; scavenges ABTS radicals in a cell-free assay (IC50 = 19.4 µM); inhibits LPS-induced NO production in RAW 264.7 macrophages (IC50 = 84.45 µM); inhibits Aβ42 self-aggregation in a cell-free assay at 20 µM
Catalog No. 50-291-1977 Supplier Cayman Chemical Supplier No. 360325 MG
A lignan; scavenges ABTS radicals in a cell-free assay (IC50 = 19.4 µM); inhibits LPS-induced NO production in RAW 264.7 macrophages (IC50 = 84.45 µM); inhibits Aβ42 self-aggregation in a cell-free assay at 20 µM