Learn More
STEMCELL Technologies EasySep™ Human HLA-DR Positive Selection and Depletion Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. NC2188053
Encompass
Isolate or deplete highly purified HLA-DR+ cells from human PBMCs or leukapheresis samples by immunomagnetic selection
This item is not returnable. View return policy
Catalog No. NC2188053 Supplier STEMCELL Technologies Supplier No. 1000980
Isolate or deplete highly purified HLA-DR+ cells from human PBMCs or leukapheresis samples by immunomagnetic selection