Learn More
World Precision Instrument Isolated, Low Noise Bioamplifier Single Channel Module
SDP
Catalog No. 502521233
Encompass
Isolated, low noise bio-amplifer
The ISO-DAM8A is a compact modular standard rack-mountable DC amplifier system. Each channel is electrically isolated from the others and from ground. No current can flow from the input terminals and electrodes. The instrument is intrinsically safe and cannot cause any electrical stimulus or shock to the preparation. In addition, ground loop noise is minimized.
Catalog No. 50-252-1233 Supplier World Precision Instrument Supplier No. 74020
Isolated, low noise bio-amplifer
The ISO-DAM8A is a compact modular standard rack-mountable DC amplifier system. Each channel is electrically isolated from the others and from ground. No current can flow from the input terminals and electrodes. The instrument is intrinsically safe and cannot cause any electrical stimulus or shock to the preparation. In addition, ground loop noise is minimized.