Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Isolation of E. Coli Chromosomal DNA
SDPSCI_ED

Catalog No. s11203 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
5 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S11203 5 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S11203 Supplier Edvotek™ Supplier No. 203
Only null left
Add to Cart
Add to Cart

Includes

instructions, Chromosomal LyphoCells™, various solutions and buffers, agarose powder, FlashBlue™ Stain.

Requires

  • Horizontal gel electrophoresis apparatus
  • DC power supply
  • Automatic micropipet with tips
  • Waterbath
  • Visualization system (white light)
  • 50 and 250mL beakers or flasks
  • 5 and 10mL pipets
  • Test tubes for DNA resuspension
  • 99–100% Isopropanol (70% Isopropyl rubbing alcohol can be substituted)
  • Distilled or deionized water

Isolate chromosomal DNA from E. Coli.

Isolation of high molecular weight chromosomal DNA is the first step in molecular cloning since it is the source of genes in cells. This experiment provides DNA Extraction LyphoCells™ and reagents for isolating chromosomal DNA from E. coli. After spooling from solution, the DNA can be dissolved and analyzed by agarose gel electrophoresis as an optional lab extension activity.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Isolating chromosomal DNA
Includes Instructions, Chromosomal LyphoCells™, Various solutions and buffers, Agarose powder, FlashBlue™ stain
Quantity 20 DNA Isolations and 5 Gels
Requires Water bath, pipet pumps or bulbs, lab glassware, distilled or deionized water, 95-100% isopropanol. For Optional Electrophoresis: Electrophoresis apparatus, power supply, automatic micropipet with tips, balance, microwave oven or hot plate, misc. labware, white light visualization system, and photodocumentation system.
Time Range 1 hour 45 minutes
Content And Storage Room temperature
Class Size 5 Lab Groups
Product Type Electrophoresis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.