Learn More
Med Vet International Level 4 Disposable Isolation Gown, Non-Sterile, Breathable Film, X-Large, 30 Count
Catalog No. NC2310132
Encompass_Preferred
Impervious Gown: Prevention™ Plus gowns offer protection against heavy fluids. Made from blue impervious breathable material for high fluid protection. Styled with a hook-and-loop neck closure, knit cuffs and waist ties. Tested against top chemo drugs. Blue, X-Large Size.Level 4
Catalog No. NC2310132 Supplier Med Vet International Supplier No. NON27457XL
Impervious Gown: Prevention™ Plus gowns offer protection against heavy fluids. Made from blue impervious breathable material for high fluid protection. Styled with a hook-and-loop neck closure, knit cuffs and waist ties. Tested against top chemo drugs. Blue, X-Large Size.Level 4