Learn More
STEMCELL Technologies Mitochondrial Isolation Kit, Size: 50 Tests
SDP
Catalog No. 502250345
Encompass
Conveniently enrich the mitochondrial or cytosolic subcellular fractions from mammalian cells or tissues using the Mitochondrial Isolation Kit. This kit offers two quick options to isolate the mitochondrial subcellular fraction and has been verified for use with western blot analysis. The first option is a reagent-based method for processing up to six samples concurrently. The second option utilizes Dounce homogenization, providing a two-fold increase in isolated mitochondria from a single sample. Both options use a microcentrifuge-based method to separate the mitochondrial fraction from the cytosolic fraction while preserving their biological activity. This separation is a valuable tool for studying apoptosis and signaling events between the two fractions. For more information on the Mitochondrial Isolation Kit, visit www.stemcell.com.
This item is not returnable. View return policy
Catalog No. 50-225-0345 Supplier STEMCELL Technologies Supplier No. 1000990
Conveniently enrich the mitochondrial or cytosolic subcellular fractions from mammalian cells or tissues using the Mitochondrial Isolation Kit. This kit offers two quick options to isolate the mitochondrial subcellular fraction and has been verified for use with western blot analysis. The first option is a reagent-based method for processing up to six samples concurrently. The second option utilizes Dounce homogenization, providing a two-fold increase in isolated mitochondria from a single sample. Both options use a microcentrifuge-based method to separate the mitochondrial fraction from the cytosolic fraction while preserving their biological activity. This separation is a valuable tool for studying apoptosis and signaling events between the two fractions. For more information on the Mitochondrial Isolation Kit, visit www.stemcell.com.