Learn More
Grainger Tripp Lite Medical-Grade Isolation Transformer, 115/230V, 1000W, 8 Outlets, 1 ea
Catalog No. 502785460
Encompass_Preferred
The medical-grade isolation transformer is designed to protect sensitive electronic equipment in patient-care vicinities and other industrial or network environments. It features an internal low-impedance toroidal transformer that provides 100% isolation from the input mains AC line, while a Faraday shield reduces cumulative leakage current to less than 80 μA. This versatile unit supports step-up, step-down, or pass-through voltage operation and provides continuous EMI/RFI noise filtering and surge protection.
  • Full CAN/ES/IEC/EN 60601-1 medical-grade listing
  • Eight C13 outlets with fuse overload protection
  • Configurable for 115V or 230V operation
  • Reduces 6000V surge test transients to 0.5V common mode
  • Rugged all-metal housing
  • Continuous common-mode noise rejection with no wearable parts
  • Includes lighted on/off switch
Catalog No. 50-278-5460 Supplier Grainger Supplier No. 198ZH5
The medical-grade isolation transformer is designed to protect sensitive electronic equipment in patient-care vicinities and other industrial or network environments. It features an internal low-impedance toroidal transformer that provides 100% isolation from the input mains AC line, while a Faraday shield reduces cumulative leakage current to less than 80 μA. This versatile unit supports step-up, step-down, or pass-through voltage operation and provides continuous EMI/RFI noise filtering and surge protection.
  • Full CAN/ES/IEC/EN 60601-1 medical-grade listing
  • Eight C13 outlets with fuse overload protection
  • Configurable for 115V or 230V operation
  • Reduces 6000V surge test transients to 0.5V common mode
  • Rugged all-metal housing
  • Continuous common-mode noise rejection with no wearable parts
  • Includes lighted on/off switch