Learn More
Waters Corp Waters Isolation Valve, Mass Spectrometry, Spare Part
Catalog No. 50792196 Shop All Waters Corporation Products
Encompass_Preferred
This isolation valve is a critical spare part for mass spectrometry systems. It is designed to separate the instrument’s source from the mass spectrometer, allowing for essential maintenance while preserving the system’s vacuum integrity.
  • Isolates the source from the mass spectrometer.
  • Enables maintenance on the source without venting the entire mass spectrometer.
  • Maintains vacuum integrity during source-related work.
  • Reconnects the source to the mass spectrometer’s vacuum system following maintenance.
  • Assists the system in achieving operational vacuum after service.
Catalog No. 50-792-196 Supplier Waters Corp Supplier No. 6062807
This isolation valve is a critical spare part for mass spectrometry systems. It is designed to separate the instrument’s source from the mass spectrometer, allowing for essential maintenance while preserving the system’s vacuum integrity.
  • Isolates the source from the mass spectrometer.
  • Enables maintenance on the source without venting the entire mass spectrometer.
  • Maintains vacuum integrity during source-related work.
  • Reconnects the source to the mass spectrometer’s vacuum system following maintenance.
  • Assists the system in achieving operational vacuum after service.