Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ Isolator™ Microbial Tubes

Catalog No. 22769667 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Tube Size:
Adult
Pediatric
Volume (Metric):
1.5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tube Size Volume (Metric)
22-769667 Adult 10 mL
22-769666 Pediatric 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-769667 Supplier Abbott™ Supplier No. 50C7

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Evacuated tube with lysing and anticoagulating agents specifically designed for collecting and isolating microorganisms. Adult and pediatric tubes available.

Benefits:

  • Blood culture system for the collection and isolation of microorganisms from the blood. The ISOLATOR™ tube is used to concentrate and deliver microbial organisms to conventional growth media for purposes of isolation, identification and susceptibility testing.
  • When compared to conventional broth systems, lysis centrifugation can reduce the time to isolation by 24 hours

 

Specifications

Product Type Microbial Tube
For Use With (Application) For collection and isolation of microorganisms from blood
For Use With (Equipment) ISOSTAT™ System Press and Rack
Quantity 25 Pk.
Storage Requirements 2°C to 40°C
Tube Size Adult
Volume (Metric) 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.