These green nitrile glovebox gloves provide reliable protection and comfort in controlled environments. Constructed from a proprietary formulation, these 23 mil thick gloves deliver chemical resistance, mechanical protection, and a soft, flexible feel. The one-piece design offers enhanced safety and convenience compared to traditional assemblies.
- One-piece design increases safety compared to glove-and-sleeve assemblies
- Proprietary nitrile formulation provides chemical resistance and mechanical protection
- Lightweight, soft, and flexible construction improves wearer comfort
- Compatible with alcohol-based cleaning practices
- Textured surface ensures a secure grip on wet and dry objects
- Inherently anti-static with a surface resistance of 1011 Ω/sq
- Compliant with 21 CFR regulations for food processing
- Dimensions: 27 in. length, 8 in. cuff diameter