Learn More
Thermo Scientific™ Isolator Wrap Sterile Settle Tryptic Soy Agar Plates
Shop All Thermo Scientific ProductsDescription
- VHP resistant packaged Settling plates (single bag and double bag) are manufactured in a clean room environment. Irradiated with test indicators. This provides the highest level of sterility assurance with no compromise of growth characteristics.
- Each lot is tested to ensure consistent, reproducible results.
- Storage Condition: 2 to 8°C.
Specifications
Specifications
| Content And Storage | 2°C to 8°C, Refrigerated |
| Media Type | Tryptic Soy Agar |
| Product Type | Isolator Settling Plate |
| For Use With (Equipment) | RABS, Isolators and Cleanrooms |
| Quantity | 10/Pk. |
| Diameter (Metric) | 85 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.