Learn More
Enzo Life Sciences Isolectin B4 (Bandeiraea simplicifolia), (FITC conjugate) (0.5mg)
SDP
Supplier:  Enzo Life Sciences ALX650001FMC05
Encompass_Preferred
Alternative name: Isolectin B4 (Griffonia simplicifolia), BSI-B4. Concentration: 1mg/ml. Formulation: Liquid. In 10mM HEPES, pH 7.5, containing 0.15M sodium chloride, 0.08% sodium azide and 0.1mM Ca2+. Source: Isolated from Griffonia (Bandeiraea) simplicifolia seeds. Specificity: Binds to α-(1,3) galactosyl galactoside epitopes. Long Term Storage: +4°C.
Catalog No. 50-200-9657