Learn More
Medchemexpress LLC Isoleucine methyl ester hydrochloride | 18598-74-8 | MFCD00038911 | >98.0% | 181.66 | C7H16ClNO2 | 100 G
SDP
Catalog No. 5000303253
Encompass_Preferred
Isoleucine methyl ester hydrochloride is the hydrochloride salt of the methyl ester of isoleucine, provided as a research-grade amino acid derivative and peptide synthesis building block. It is used as an intermediate in peptide assembly and organic synthesis, and is suitable for laboratory and synthetic applications.
  • High purity (≥98.0%).
  • White to off-white solid, easy to handle.
  • Suitable as a building block for peptide synthesis.
  • Stable when stored sealed at 4°C; in solution, store at -80°C for long-term preservation.
  • Well characterized with CAS 18598-74-8 and established identifiers for traceability.
Catalog No. 50-003-03253 Supplier Medchemexpress LLC Supplier No. HYW016423100G
Isoleucine methyl ester hydrochloride is the hydrochloride salt of the methyl ester of isoleucine, provided as a research-grade amino acid derivative and peptide synthesis building block. It is used as an intermediate in peptide assembly and organic synthesis, and is suitable for laboratory and synthetic applications.
  • High purity (≥98.0%).
  • White to off-white solid, easy to handle.
  • Suitable as a building block for peptide synthesis.
  • Stable when stored sealed at 4°C; in solution, store at -80°C for long-term preservation.
  • Well characterized with CAS 18598-74-8 and established identifiers for traceability.