Learn More
Selleck Chemical LLC Isoliquiritigenin 10mg 961-29-5
SDP
Supplier:  Selleck Chemical LLC S240410MG
Encompass_Preferred
Isoliquiritigenin, an anti-tumor flavonoid from the root of Glycyrrhiza glabra, inhibits aldose reductase with an IC50 of 320 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-685-0