Learn More
Cayman Chemical ISOLIQUIRITIN 10MG
SDP
Catalog No. 502992529
Encompass_Preferred
A flavonoid with diverse biological activities; inhibits LPS-induced nitrite and PGE2 production in RAW 264.7 cells at 1.6 µM; inhibits tube formation by isolated rat aortic endothelial cells from 5-500 UG/ml; reduces carmine content and pouch fluid weight in a mouse model of adjuvant-induced pouch granuloma formation (ED50s = 1.46 and 0.771 mg/kg, respectively); increases cortical, hippocampal, and hypothalamic 5-HT and norepinephrine levels and decreases immobility time in the forced swim and tail suspension tests in mice from 10-40 mg/kg
Catalog No. 50-299-2529 Supplier Cayman Chemical Supplier No. 3027010 MG
A flavonoid with diverse biological activities; inhibits LPS-induced nitrite and PGE2 production in RAW 264.7 cells at 1.6 µM; inhibits tube formation by isolated rat aortic endothelial cells from 5-500 UG/ml; reduces carmine content and pouch fluid weight in a mouse model of adjuvant-induced pouch granuloma formation (ED50s = 1.46 and 0.771 mg/kg, respectively); increases cortical, hippocampal, and hypothalamic 5-HT and norepinephrine levels and decreases immobility time in the forced swim and tail suspension tests in mice from 10-40 mg/kg