Learn More
Selleck Chemical LLC Isoliquiritin apioside S0923-5MG
SDP
Catalog No. 5000042901
Encompass_Preferred
Isoliquiritin apioside (ISLA ILA) a component isolated from Glycyrrhizae radix rhizome (GR) significantly decreases PMA-induced increases in matrix metalloproteinase (MMP) activities and suppresses PMA-induced activation of mitogen-activated protein kinase (MAPK) and NF- B Isoliquiritin apioside possesses anti-metastatic and anti-angiogenic abilities in malignant cancer cells and ECs with no cytotoxicity
Catalog No. 50-000-42901 Supplier Selleck Chemical LLC Supplier No. S09235MG
Isoliquiritin apioside (ISLA ILA) a component isolated from Glycyrrhizae radix rhizome (GR) significantly decreases PMA-induced increases in matrix metalloproteinase (MMP) activities and suppresses PMA-induced activation of mitogen-activated protein kinase (MAPK) and NF- B Isoliquiritin apioside possesses anti-metastatic and anti-angiogenic abilities in malignant cancer cells and ECs with no cytotoxicity