Learn More
TARGETMOL CHEMICALS INC ISOLIQUIRITIN APIOSIDE 5MG
SDP
Catalog No. 502580102
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Isoliquiritin apioside isolated from Glycyrrhizae radix rhizome significantly decreases PMA-induced increases in MMP9 activities and suppresses PMA-induced activation of MAPK and NF-(kappa)B. Isoliquiritin apioside auppresseses invasiveness and angiogenesis of cancer cells and endothelial cells. Isoliquiritin apioside has marked potential to combat oxidative stress-induced genotoxicity. purity: 99%
Catalog No. 50-258-0102 Supplier TARGETMOL CHEMICALS INC Supplier No. T38605MG
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Isoliquiritin apioside isolated from Glycyrrhizae radix rhizome significantly decreases PMA-induced increases in MMP9 activities and suppresses PMA-induced activation of MAPK and NF-(kappa)B. Isoliquiritin apioside auppresseses invasiveness and angiogenesis of cancer cells and endothelial cells. Isoliquiritin apioside has marked potential to combat oxidative stress-induced genotoxicity. purity: 99%