Learn More
TARGETMOL CHEMICALS INC Isolithocholic Acid 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T357921MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Isolithocholic Acid (3-Epilithocholic acid) (beta-Lithocholic acid) is an isomer of Lithocholic acid. Isolithocholic Acid, a bile acid, is formed by microbial metabolism of Lithocholic acid or Lithocholic acid 3alpha-sulfate. Purity 99.4%
Catalog No. 50-287-6683