Learn More
Medchemexpress LLC HY-N7363 5mg Medchemexpress, Isolongifolene CAS:1135-66-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN73635MG
Encompass_Preferred
Medchemexpress, HY-N7363 5mg Isolongifolene CAS:1135-66-6 Isolongifolene ((-)-Isolongifolene) is a tricyclic sesquiterpene isolated from Murraya koenigii. Isolongifolene attenuates Rotenone-induced oxidative stress, mitochondrial dysfunction and apoptosis through the regulation of P13K/AKT/GSK-3β signaling pathways. Isolongifolene has antioxidant, anti-inflammatory, anticancer and neuroprotective properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2992