Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (-)-Isolongifolene

Catalog No. 589241ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-924-1ML 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 58-924-1ML Supplier Sigma Aldrich Supplier No. 589241ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(-)-Isolongifolene is a sesquiterpene that can be found in pine cone and Japanese cedar essential oils.(-)-Isolongifolene may be used in the preparation of (-)-isolongifolenone, a potent insect repellent.This (-)-isolongifolene has an enantiomeric purity of 93%; it is thus the highest purity material ever offered commercially; the double bond can be epoxidized

Specifications

CAS 99769-19-4
Melting Point 205°C to 208°C
Molecular Formula C8H9BO4
Refractive Index n20/D 1.499
Linear Formula CH3O2CC6H4B(OH)2
MDL Number MFCD02093046
Quantity 5 g
Synonym Methyl 3-boronobenzoate
Molecular Weight (g/mol) 179.97 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.