Learn More
Biotage ISOLUTE C18 500 mg/6 mL (Tabless), 30/pk
SDP
Catalog No. NC1915361
Encompass
ISOLUTE C18 is the most commonly used C18 sorbent for the extraction of acidic, neutral and basic compounds from aqueous sample matrices. The sorbent is not end-capped, and secondary silanol interactions can be exploited to enhance extract purity and method robustness for basic analytes.
Catalog No. NC1915361 Supplier Biotage Supplier No. 2200050CG
ISOLUTE C18 is the most commonly used C18 sorbent for the extraction of acidic, neutral and basic compounds from aqueous sample matrices. The sorbent is not end-capped, and secondary silanol interactions can be exploited to enhance extract purity and method robustness for basic analytes.