Learn More
Biotage ISOLUTE C8(EC) | 500mg/6mL
SDP
Catalog No. NC1775082
Encompass
ISOLUTE C8(EC) is a medium retentive, non-polar sorbent for extraction of a range of analytes from aqueous samples using a non-polar retention mechanism.
Catalog No. NC1775082 Supplier Biotage Supplier No. 2910050C
ISOLUTE C8(EC) is a medium retentive, non-polar sorbent for extraction of a range of analytes from aqueous samples using a non-polar retention mechanism.