Learn More
Biotage ISOLUTE SI 10 g/70 mL, 16/pk
SDP
Catalog No. NC1923262
Encompass
ISOLUTE SI is an unbonded silica sorbent. It is used to isolate polar analytes from non-polar matrices using hydrogen bonding interactions. The moisture content of ISOLUTE SI is controlled to 7% (+/- 1%) to aid method robustness.
Catalog No. NC1923262 Supplier Biotage Supplier No. 4601000F
ISOLUTE SI is an unbonded silica sorbent. It is used to isolate polar analytes from non-polar matrices using hydrogen bonding interactions. The moisture content of ISOLUTE SI is controlled to 7% (+/- 1%) to aid method robustness.