Learn More
CTL Scientific Supply Corp ISOLYMPH PK OF 6 X 500 ML
SDP
Supplier:  CTL Scientific Supply Corp 759050
Encompass_Preferred
Isolymph is used for Lymphocyte separation in laboratory research. Density is 1.077 +/- 0.001 g/ml
Catalog No. 50-300-403