Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isomalt Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015045 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5045 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5045 Supplier MilliporeSigma Supelco Supplier No. PHR17691G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isomalt is a disaccharide alcohol that is commonly used as an excipient in pharmaceutical formulations because of its beneficial characteristics such as the taste, mouth feel, low calorie content, acariogenicity, suitability to diabetics, high stability and low hygroscopicity.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 64519-82-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C12H24O11
Synonym Isomalt; 6-O-alpha-D-Glucopyranosyl-D-glucitol mixed with 1-O-alpha-D-glucopyranosyl-D-mannitol
Type Certified Reference Material
Quantity 1 g
Formula Weight 344.31
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.