Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isomaltose 97.0+%, TCI America™
SDP

Catalog No. I02311G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I02311G 1 g
I0231100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I02311G Supplier TCI America Supplier No. I02311G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 499-40-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.297
MDL Number MFCD00065373
InChI Key DLRVVLDZNNYCBX-MYBWVJKRSA-N
Synonym isomaltose
PubChem CID 133612057
IUPAC Name (2S,3S,4R,5R,6S)-6-[[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxane-2,3,4,5-tetrol
SMILES C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)O)O)O)O)O)O)O)O

Specifications

Color White
Quantity 1 g
Formula Weight 342.30
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Isomaltose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.