Learn More
Medchemexpress LLC Isomaltotriose | 3371-50-4 | 504.44 | 1 ML
SDP
Catalog No. 5000416839
Encompass_Preferred
Isomaltotriose is a sugar derived from enzymic hydrolyzates of dextran from *Leuconostoc mesenteroides* NRRL B-512. This high-purity product (98.0%) is suitable for various research applications. It is supplied as a 10 mM solution in 1 mL of water, ready for use, and is also highly soluble in H₂O and PBS.
  • Derived from enzymic hydrolyzates of dextran
  • High purity (98.0%)
  • Soluble in water at concentrations ≥ 100 mg/mL
  • Soluble in PBS (100 mg/mL) with sonication
  • Supplied as a 10 mM solution in 1 mL of water
Catalog No. 50-004-16839 Supplier Medchemexpress LLC Supplier No. HYN0913A10MM/1ML
Isomaltotriose is a sugar derived from enzymic hydrolyzates of dextran from *Leuconostoc mesenteroides* NRRL B-512. This high-purity product (98.0%) is suitable for various research applications. It is supplied as a 10 mM solution in 1 mL of water, ready for use, and is also highly soluble in H₂O and PBS.
  • Derived from enzymic hydrolyzates of dextran
  • High purity (98.0%)
  • Soluble in water at concentrations ≥ 100 mg/mL
  • Soluble in PBS (100 mg/mL) with sonication
  • Supplied as a 10 mM solution in 1 mL of water