Learn More
Medchemexpress LLC Isomaltotriose | 3371-50-4 | 504.44 | 50 MG
SDP
Catalog No. 5000415312
Encompass_Preferred
Isomaltotriose is a sugar derived from enzymic hydrolyzates of the dextran from Leuconostoc mesenteroides NRRL B-512. It is intended for research use only.
  • Derived from enzymatic hydrolysis of dextran.
  • For research use only.
  • High purity (98.0%).
  • Available with supporting documentation (Data Sheet, COA, SDS, Handling Instructions).
  • Stable under recommended storage conditions.
Catalog No. 50-004-15312 Supplier Medchemexpress LLC Supplier No. HYN0913A50MG
Isomaltotriose is a sugar derived from enzymic hydrolyzates of the dextran from Leuconostoc mesenteroides NRRL B-512. It is intended for research use only.
  • Derived from enzymatic hydrolysis of dextran.
  • For research use only.
  • High purity (98.0%).
  • Available with supporting documentation (Data Sheet, COA, SDS, Handling Instructions).
  • Stable under recommended storage conditions.