Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Isomaltulose hydrate, 98%

Catalog No. AC225951000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC225951000 100 g Glass bottle
AC225950250 25 g Glass bottle
AC225955000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC225951000 Supplier Thermo Scientific Chemicals Supplier No. 225951000
Only null left
Add to Cart

CAS: 343336-76-5 | C12H22O11 | 342.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 343336-76-5
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD11100373
InChI Key PVXPPJIGRGXGCY-VVLSOAGWNA-N
Synonym 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2r,3s,4s,5r-3,4,5-trihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl methoxy tetrahydro-2h-pyran-3,4,5-triol hydrate
PubChem CID 126970043
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[[(2R,3S,4R,5R)-3,4,5-trihydroxy-5-(hydroxymethyl)oxolan-2-yl]methoxy]oxane-3,4,5-triol;hydrate
SMILES OC[C@H]1O[C@H](OC[C@H]2O[C@@](O)(CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 122.0°C to 124.0°C
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 100 g
Specific Rotation + 100.00
Formula Weight 342.3
Percent Purity 98%
Chemical Name or Material Isomaltulose hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.