Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isomannide (1,4:3,6-dianhydromannitol) | 641-74-7 | MFCD10698761 | 99.9% | 146.14 g/mol | C6H10O4 | 100 G
SDP

Catalog No. 5000303227
Encompass_Preferred

Isomannide (1,4:3,6-dianhydromannitol) is a small-molecule intermediate and biochemical reagent used in synthetic chemistry and biological research. It is supplied as a powder with high reported purity and strong DMSO solubility, and is commonly employed as a drug intermediate and in studies of HCV infection and KLK5/KLK7-related biology.

  • Small-molecule intermediate for biochemical and synthetic research.
  • Molecular formula C6H10O4; molecular weight 146.14 g/mol.
  • Provided as a powder with high reported purity.
  • Soluble in DMSO at ≥100 mg/mL.
  • Used as a drug intermediate and in HCV and KLK5/KLK7 research.
  • Storage: powder 4°C for 2 years or -20°C for 3 years; in solvent -80°C for 6 months.

Catalog No. 50-003-03227 Supplier Medchemexpress LLC Supplier No. HYW013827100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isomannide (1,4:3,6-dianhydromannitol) is a small-molecule intermediate and biochemical reagent used in synthetic chemistry and biological research. It is supplied as a powder with high reported purity and strong DMSO solubility, and is commonly employed as a drug intermediate and in studies of HCV infection and KLK5/KLK7-related biology.

  • Small-molecule intermediate for biochemical and synthetic research.
  • Molecular formula C6H10O4; molecular weight 146.14 g/mol.
  • Provided as a powder with high reported purity.
  • Soluble in DMSO at ≥100 mg/mL.
  • Used as a drug intermediate and in HCV and KLK5/KLK7 research.
  • Storage: powder 4°C for 2 years or -20°C for 3 years; in solvent -80°C for 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.