Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isomannide (1,4:3,6-dianhydromannitol) | 641-74-7 | MFCD10698761 | 99.9% | 146.14 g/mol | C6H10O4 | 25 G
SDP

Catalog No. 5000303512
Encompass_Preferred

Isomannide (1,4:3,6-dianhydromannitol) is a small-molecule intermediate used in biochemical research and synthetic chemistry. Supplied as a powder with high reported purity, it is applied as a building block, reagent, or derivative scaffold in studies including viral infection models and protease-related pathways. For research use only.

  • High purity (99.93%).
  • Molecular formula C6H10O4; molecular weight 146.14 g/mol.
  • Powder form for easy handling and weighing.
  • Soluble in DMSO at ≥100 mg/mL (saturation not specified).
  • Storage: powder stable at -20°C for up to 3 years; in solvent, store at -80°C for best stability.
  • Intended use as an intermediate, reagent, or drug-derivative building block in research.

Catalog No. 50-003-03512 Supplier Medchemexpress LLC Supplier No. 25GHYW013827
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isomannide (1,4:3,6-dianhydromannitol) is a small-molecule intermediate used in biochemical research and synthetic chemistry. Supplied as a powder with high reported purity, it is applied as a building block, reagent, or derivative scaffold in studies including viral infection models and protease-related pathways. For research use only.

  • High purity (99.93%).
  • Molecular formula C6H10O4; molecular weight 146.14 g/mol.
  • Powder form for easy handling and weighing.
  • Soluble in DMSO at ≥100 mg/mL (saturation not specified).
  • Storage: powder stable at -20°C for up to 3 years; in solvent, store at -80°C for best stability.
  • Intended use as an intermediate, reagent, or drug-derivative building block in research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.