Learn More
Medchemexpress LLC Isomannide (1,4:3,6-dianhydromannitol) | 641-74-7 | MFCD10698761 | 99.9% | 500g
SDP
Catalog No. 5000302282
Encompass_Preferred
Isomannide (1 4 3 6-Dianhydromannitol) is a common intermediate compound and derivative of Isosorbide (HY-B1469) Some derivatives of isomannide have potent inhibitory effects on KLK5 and KLK7 Isomannide can be used in the research of HCV infection and KLK5/KLK7-related diseases[1][2][3]
Catalog No. 50-003-02282 Supplier Medchemexpress LLC Supplier No. HYW013827500G
Isomannide (1 4 3 6-Dianhydromannitol) is a common intermediate compound and derivative of Isosorbide (HY-B1469) Some derivatives of isomannide have potent inhibitory effects on KLK5 and KLK7 Isomannide can be used in the research of HCV infection and KLK5/KLK7-related diseases[1][2][3]