Learn More
TARGETMOL CHEMICALS INC ISOMER-GAT 107 5MG
SDP
Catalog No. 502580616
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. isomer-GAT 107 ((-)GAT 107) is an inactive isomer of (+)GAT 107. purity: 99%
Catalog No. 50-258-0616 Supplier TARGETMOL CHEMICALS INC Supplier No. T27404L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. isomer-GAT 107 ((-)GAT 107) is an inactive isomer of (+)GAT 107. purity: 99%