Learn More
Perkin Elmer US LLC IsoMist Cyclonic Spray Chamber Inlet System Kit for NexION 300/350
Supplier:  Perkin Elmer US LLC N8141426
Encompass_Preferred
The IsoMist Programmable Temperature Spray Chamber lets you select any temperature between -10 °C and +60 °C in 1 °C increments to provide the optimum conditions for any application. The rapid response of the Peltier device allows a spray chamber temperature of -5 °C to be reached within 15 minutes. The spray chamber temperature can be monitored through a temperature versus time plot on your PC screen. And, for regulatory compliance, the temperature versus time data can be saved with your results. If these features are not required, once the temperature has been programmed, the IsoMist can be run in stand-alone mode without a PC connection. Temperature range: -10 °C to +60 °C Temperature set in 1 °C increments Temperature stability: +/- 0.1 °C Cools to -5 °C within 15 minutes Wireless Bluetooth or USB connection to PC PC operating system: Windows 2000 or XP Weight: 2 kg Dimensions: 195 x 100 x 120 mm Input Voltage: 100 - 250 VAC Input Frequency: 47 - 63 Hz Maximum Current: 2 A Maximu
Catalog No. 50-211-7227
Add to Cart
Request a Quote