Learn More
Glass Expansion IsoMist XR Basic Kit ROL
SDP
Catalog No. 501907622
Encompass_Preferred
IsoMist XR Basic Kit ROL. Compatible with all ICP-OES and ICP-MS. This IsoMist XR is programmable and temperature-controlled, with a glass cyclonic spray chamber now featuring an improved thermodynamic design that provides an extended temperature range of -25°C to +80°C. The temperature is settable in 1°C increments and can maintain accuracy of + or – 0.1°C. Proven performance results are, improved long-term stability and precision, no dilution for volatile organic solvents, reduced oxide interferences (ICP-MS), enhanced sensitivity and accuracy, lower detection limits, faster washout, and reduced hazardous waste/disposal fees.Replacement parts include 502703176, GAZ-04-90, and 501907304.The IsoMist XR comes standard with a borosilicate glass spray chamber. Optional quartz or PFA HF-resistant spray chambers can be ordered separately.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7622 Supplier Glass Expansion Supplier No. KT1024XR
IsoMist XR Basic Kit ROL. Compatible with all ICP-OES and ICP-MS. This IsoMist XR is programmable and temperature-controlled, with a glass cyclonic spray chamber now featuring an improved thermodynamic design that provides an extended temperature range of -25°C to +80°C. The temperature is settable in 1°C increments and can maintain accuracy of + or – 0.1°C. Proven performance results are, improved long-term stability and precision, no dilution for volatile organic solvents, reduced oxide interferences (ICP-MS), enhanced sensitivity and accuracy, lower detection limits, faster washout, and reduced hazardous waste/disposal fees.Replacement parts include 502703176, GAZ-04-90, and 501907304.The IsoMist XR comes standard with a borosilicate glass spray chamber. Optional quartz or PFA HF-resistant spray chambers can be ordered separately.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001