Learn More
Perkin Elmer US LLC IsoMist XR Quartz Spray Chamber for Optima 4x00/5x00/7x00 DV/8x00
Supplier:  Perkin Elmer US LLC N0780761
Encompass_Preferred
The IsoMist Programmable Temperature Controlled Spray Chamber provides the benefits of a temperature controlled ICP sample introduction system in a compact, convenient package. The temperature is electronically controlled using a powerful inbuilt Peltier device. You can select any temperature between -10°C and +60°C in 1°C increments to provide the optimum conditions for any application. The rapid response of the Peltier device allows a spray chamber temperature of -5°C to be reached within 15 minutes. The IsoMist XR programmable temperature controlled cyclonic spray chamber now features an improved thermodynamic design providing an extended temperature range and faster cool-down, so your ICP is ready to go sooner. IsoMist XR has a peltier unit with a range of -25°C to +80°C. Features and Benefits include: Improve Analytical Stability Reduce Oxide Formation Perfect for Volatile Organics Constant Temperature Improves Analytical Stability Enhance Sensitivity with Elevated Sample Introd
Catalog No. 50-210-5382
Add to Cart
Request a Quote