Learn More
Medchemexpress LLC Isomogroside V | 1126032-65-2 | 10 MG
SDP
Catalog No. 5000419969
Encompass_Preferred
Isomogroside V is a sweetener extracted from the plant *Siraitia grosvenorii (Swingle)*. This product is intended for research use only and is not for sale to patients.
  • Extracted from *Siraitia grosvenorii (Swingle)*
  • Suitable for research use only
  • Appears as a solid
  • Color is white to off-white
  • Classified as a triterpene
Catalog No. 50-004-19969 Supplier Medchemexpress LLC Supplier No. HYN681510MG
Isomogroside V is a sweetener extracted from the plant *Siraitia grosvenorii (Swingle)*. This product is intended for research use only and is not for sale to patients.
  • Extracted from *Siraitia grosvenorii (Swingle)*
  • Suitable for research use only
  • Appears as a solid
  • Color is white to off-white
  • Classified as a triterpene