Learn More
TARGETMOL CHEMICALS INC Isomucronulatol 7-O-glucoside
SDP
Catalog No. 502877267
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. Isomucronulatol 7-O-glucoside, a natural product, has a molecular formula of C21H22O11. It is known for its glycosidic linkage between isomucronulatol and glucose, which contributes to its biological activity and potential therapeutic applications. This compound can be isolated from various plant species and has been studied for its pharmacological properties. Purity 99.33%
Catalog No. 50-287-7267 Supplier TARGETMOL CHEMICALS INC Supplier No. TN17875MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. Isomucronulatol 7-O-glucoside, a natural product, has a molecular formula of C21H22O11. It is known for its glycosidic linkage between isomucronulatol and glucose, which contributes to its biological activity and potential therapeutic applications. This compound can be isolated from various plant species and has been studied for its pharmacological properties. Purity 99.33%