Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoniazid, ≥99% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111014577 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4577 5 g
11-101-4576 50 g
11-101-4575 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-4577 Supplier MilliporeSigma Supelco Supplier No. I33775G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical structure: pyridine

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Poly Bottle
CAS 54-85-3
Physical Form Neat
Linear Formula C6H7N3 O
MDL Number MFCD00006426
Melting Point 171°C to 173°C
Synonym 4-Pyridinecarboxylic acid hydrazide; INH; Isonicotinic acid hydrazide; Isonicotinic hydrazide
Type Analytical Standard
Quantity 5 g
Formula Weight 137.14
Percent Purity ≥99% (TLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.