Learn More
Medchemexpress LLC Isoniazid-d4 | 774596-24-6 | 99.9% | 5 MG
SDP
Catalog No. 5000406712
Encompass_Preferred
Isoniazid-d4 is the deuterium-labeled Isoniazid. Isoniazid (INH) is a proagent that requires activation by the bacterial catalase-peroxidase enzyme KatG. It is bactericidal to rapidly dividing mycobacteria and exhibits anti-tuberculostatic activity.
  • Can be utilized as a tracer
  • Can serve as an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS
  • Deuteration has potential to influence pharmacokinetic and metabolic profiles of drugs
  • Suitable for research use only
Catalog No. 50-004-06712 Supplier Medchemexpress LLC Supplier No. HYB0329S5MG
Isoniazid-d4 is the deuterium-labeled Isoniazid. Isoniazid (INH) is a proagent that requires activation by the bacterial catalase-peroxidase enzyme KatG. It is bactericidal to rapidly dividing mycobacteria and exhibits anti-tuberculostatic activity.
  • Can be utilized as a tracer
  • Can serve as an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS
  • Deuteration has potential to influence pharmacokinetic and metabolic profiles of drugs
  • Suitable for research use only