Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoniazid, G-Biosciences
SCI_ED

Catalog No. S139753
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139753 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139753 Supplier G-Biosciences Supplier No. RC1042
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isoniazid is a prodrug derivative of nicotinic acid that is used as an antibiotic compound against Mycobactrium tuberculosis, M. bovis, and other mycobacteria.

This prodrug is transformed by catalase-peroxidases, where its active form inhibits an enoyl-reductase (InhA). Inhibition of InhA compromises bacterial cell walls by preventing mycolic acid synthesis, disrupting fluid homeostasis and leading to cell death.

Chemical Identifiers

Molecular Weight (g/mol) 137.14

Specifications

Physical Form Powder
Quantity 100 g
Product Line Gibco
Percent Purity 0.95
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.