Learn More
Medchemexpress LLC Isonicotinamide-d4 | 1219799-40-2 | 99.9% | 126.15 | 5 MG
SDP
Catalog No. 5000454876
Encompass_Preferred
Isonicotinamide-d4 is the deuterium labeled Isonicotinamide.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-54876 Supplier Medchemexpress LLC Supplier No. HYW012843S5MG
Isonicotinamide-d4 is the deuterium labeled Isonicotinamide.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.