Learn More
Medchemexpress LLC Isonicotinamide-d4 | 1219799-40-2 | 99.91% | 126.15 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW012843S
Encompass_Preferred
Isonicotinamide-d4 is a deuterium-labeled form of Isonicotinamide. It is commonly used as a tracer and an internal standard for quantitative analysis through techniques like NMR, GC-MS, or LC-MS. Incorporating stable heavy isotopes into drug molecules aids in quantitation during drug development, and deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Deuterium-labeled for enhanced stability and tracking
  • Can affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-52548