Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isonicotinamide (Standard) | 1453-82-3 | MFCD00006432 | 99.75% | 122.12 | 250 MG
SDP

Catalog No. 5000366378
Encompass_Preferred

Isonicotinamide (Standard) is the analytical standard of Isonicotinamide, intended for research and analytical applications. It functions as a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It is suitable for a variety of laboratory uses, including reference for assays and various research experiments.

  • Application: The compound is an analytical standard used as a reference standard for assays. It is commonly employed in qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS.
  • Appearance: Solid, white to off-white in color.
  • Shipping: Room temperature in the continental US; may vary elsewhere.
  • Storage: Store under the recommended conditions specified in the Certificate of Analysis.

Catalog No. 50-003-66378 Supplier Medchemexpress LLC Supplier No. 250MGHYW012843R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isonicotinamide (Standard) is the analytical standard of Isonicotinamide, intended for research and analytical applications. It functions as a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It is suitable for a variety of laboratory uses, including reference for assays and various research experiments.

  • Application: The compound is an analytical standard used as a reference standard for assays. It is commonly employed in qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS.
  • Appearance: Solid, white to off-white in color.
  • Shipping: Room temperature in the continental US; may vary elsewhere.
  • Storage: Store under the recommended conditions specified in the Certificate of Analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.