Learn More
Selleck Chemical LLC Isonicotinic acid
SDP
Catalog No. 5000004265
Encompass_Preferred
Isonicotinic acid (Isoniazid 4-pyridinecarboxylic acid p-pyridinecarboxylic acid 4-Picolinic acid) is a metabolite of isoniazid It is an isomer of nicotinic acid
Catalog No. 50-000-04265 Supplier Selleck Chemical LLC Supplier No. S601425MG
Add to Cart
Add to Cart
Isonicotinic acid (Isoniazid 4-pyridinecarboxylic acid p-pyridinecarboxylic acid 4-Picolinic acid) is a metabolite of isoniazid It is an isomer of nicotinic acid