Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isonicotinic acid-d4 | 53907-55-4 | 98.1% | 127.13 | C6HD4NO2 | 10 MG
SDP

Catalog No. 5000297604
Encompass_Preferred

Isonicotinic acid-d4 is the deuterium-labeled analog of isonicotinic acid, provided as an analytical standard for research and quantitative mass spectrometry. It is used as a stable isotope internal standard or tracer in metabolic and analytical studies, supplied in small laboratory pack sizes such as 10 MG.

  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity suitable for analytical applications (≈98.1%).
  • CAS number 53907-55-4 for unambiguous identification.
  • Molecular weight 127.13 g/mol for calculation and method development.
  • Available in small pack sizes, for example 10 MG, for laboratory use.
  • Store sealed and protected from moisture; in solvent: -80°C for long-term storage, -20°C for short-term storage.

Catalog No. 50-002-97604 Supplier Medchemexpress LLC Supplier No. HYI0736S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isonicotinic acid-d4 is the deuterium-labeled analog of isonicotinic acid, provided as an analytical standard for research and quantitative mass spectrometry. It is used as a stable isotope internal standard or tracer in metabolic and analytical studies, supplied in small laboratory pack sizes such as 10 MG.

  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity suitable for analytical applications (≈98.1%).
  • CAS number 53907-55-4 for unambiguous identification.
  • Molecular weight 127.13 g/mol for calculation and method development.
  • Available in small pack sizes, for example 10 MG, for laboratory use.
  • Store sealed and protected from moisture; in solvent: -80°C for long-term storage, -20°C for short-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.